N-(tert-Butoxycarbonyl)-2-(2-aminoethoxy)ethylamine 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (2-(2-aminoethoxy)ethyl)carbamate, N-Boc-2-(2-aminoethoxy)ethylamine, Amino-PEG2-NH-Boc, [2-(2-Aminoethoxy)ethyl]carbamic acid tert-Butyl ester, tert-Butyl [2-(2-Aminoethoxy)ethyl]carbamate

A108086-10G A108086-250MG A108086-25G A108086-1G A108086-5G A108086-100G
103797-292EA 91 USD
103797-292 103823-066 76982-546 76982-548 76982-550 77309-908 77309-910 77884-981
N-(tert-Butoxycarbonyl)-2-(2-aminoethoxy)ethylamine 97%
N-(tert-Butoxycarbonyl)-2-(2-aminoethoxy)ethylamine

Precaución: for Research Use Only

Formula: C₉H₂₀N₂O₃
Flash Pt: 148.013 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12031501
CAS Number: 127828-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR