tert-Butyl (2-(2-(2-hydroxyethoxy)ethoxy)ethyl)carbamate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-NH-PEG3
A163205-100MG
A163205-250MG
A163205-25G
A163205-1G
A163205-10G
103805-696EA
9
USD
103805-696
103843-954
77011-302
77011-304
77011-306
77320-042
77567-380
tert-Butyl (2-(2-(2-hydroxyethoxy)ethoxy)ethyl)carbamate 97%
tert-Butyl (2-(2-(2-hydroxyethoxy)ethoxy)ethyl)carbamate
Precaución: For research use only.
Formula:
C₁₁H₂₃NO₅ MW: 249.3 g/mol Boiling Pt: 374.6 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD07357496 CAS Number: 139115-92-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...