N-Boc-PEG2-bromide 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (2-(2-(2-bromoethoxy)ethoxy)ethyl)carbamate

A185299-1G A185299-5G A185299-100MG A185299-250MG
103815-842EA 47 USD
103815-842 103846-408 77084-048 77084-050 77164-032 77164-034
N-Boc-PEG2-bromide 97%
N-Boc-PEG2-bromide

Precaución: for Research Use Only

Formula: C₁₁H₂₂BrNO₄
Storage Temperature: Refrigerator
MDL Number: MFCD07369162
CAS Number: 165963-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR