t-Boc-N-amido-PEG4-alcohol 97%

Supplier: Ambeed
Warning

Synonyms: N-Boc-PEG4-alcohol, tert-Butyl (2-(2-(2-(2-hydroxyethoxy)ethoxy)ethoxy)ethyl)carbamate, Boc-NH-PEG4

A185146-100MG A185146-250MG A185146-1G A185146-5G A185146-25G A185146-100G
103815-822EA 23 USD
103815-822 103846-390 76721-006 76721-008 77084-024 77084-026 77600-494 77667-544
t-Boc-N-amido-PEG4-alcohol 97%
t-Boc-N-amido-PEG4-alcohol

Precaución: for Research Use Only

Formula: C₁₃H₂₇NO₆
Boiling Pt: 414.035 °C (760 mm Hg)
Flash Pt: 204.201 °C
Storage Temperature: Freezer
MDL Number: MFCD07779195
CAS Number: 106984-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR