Boc-Amino-PEG3-Amine 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (2-(2-(2-(2-aminoethoxy)ethoxy)ethoxy)ethyl)carbamate, NH2-PEG3-C2-NH-Boc

A426582-1G A426582-5G A426582-100MG A426582-250MG A426582-25G
103827-474EA 20 USD
103827-474 103838-526 76842-180 76842-182 76990-482 76990-484 76990-486
Boc-Amino-PEG3-Amine 95%
Boc-Amino-PEG3-Amine

Precaución: for Research Use Only

Formula: C₁₃H₂₈N₂O₅
MW: 292.38 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16619220
CAS Number: 101187-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR