t-Boc-N-Amido-PEG5-Amine 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (17-amino-3,6,9,12,15-pentaoxaheptadecyl)carbamate

A193534-5G A193534-100MG A193534-250MG A193534-1G
103816-552EA 139 USD
103816-552 103847-168 77085-070 77139-132 77139-134 77302-346
t-Boc-N-Amido-PEG5-Amine 95%
t-Boc-N-Amido-PEG5-Amine

Precaución: for Research Use Only

Formula: C₁₇H₃₆N₂O₇
Storage Temperature: Refrigerator
MDL Number: MFCD22056313
CAS Number: 189209-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR