t-Boc-N-Amido-PEG4-Amine 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (14-amino-3,6,9,12-tetraoxatetradecyl)carbamate, Boc-NH-PEG4-CH2CH2NH2

A198029-100MG A198029-250MG A198029-25G
103799-470EA 71 USD
103799-470 103816-986 103847-614 76873-300 76873-302 77567-858
t-Boc-N-Amido-PEG4-Amine 95%
t-Boc-N-Amido-PEG4-Amine

Precaución: For research use only.

Formula: C₁₅H₃₂N₂O₆
Storage Temperature: Refrigerator
MDL Number: MFCD16619339
CAS Number: 811442-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR