Decane-1,10-diamine, N-BOC protected 95%

Supplier: Ambeed
Warning

A509282-1G A509282-5G A509282-250MG A509282-25G
103828-368EA 14 USD
103828-368 103839-468 76843-456 76843-458 77197-094 77747-049
Decane-1,10-diamine, N-BOC protected 95%
Decane-1,10-diamine, N-BOC protected

Precaución: for Research Use Only

Formula: C₁₅H₃₂N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD02094501
CAS Number: 216961-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR