t-Butyl 1,3-dihydroxypropan-2-ylcarbamate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (1,3-dihydroxypropan-2-yl)carbamate, N-Boc-serinol

A231286-100G A231286-10G A231286-250MG A231286-25G A231286-1G A231286-5G A231286-500G
103810-768EA 6 USD
103810-768 103850-516 76874-246 76874-248 76874-250 76874-252 77089-398 77089-400 77672-052
t-Butyl 1,3-dihydroxypropan-2-ylcarbamate 97%
t-Butyl 1,3-dihydroxypropan-2-ylcarbamate

Precaución: for Research Use Only

Formula: C₈H₁₇NO₄
MW: 191.225 g/mol
Melting Pt: 85…89 °C
Flash Pt: 173.35 °C
Storage Temperature: Ambient
MDL Number: MFCD00270213
CAS Number: 125414-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR