tert-Butyl (1-(hydroxymethyl)cyclopropyl)carbamate 98%

Supplier: Ambeed
Warning

Synonyms: [1-(Hydroxymethyl)cyclopropyl]carbamic acid tert-Butyl ester, [1-(Boc-amino)cyclopropyl]methanol, tert-Butyl [1-(Hydroxymethyl)cyclopropyl]carbamate, [1-(tert-Butoxycarbonylamino)cyclopropyl]methanol

A170012-100G A170012-100MG A170012-10G A170012-250MG A170012-1G A170012-25G A170012-5G
103806-436EA 8 USD
103806-436 103844-702 76720-476 76720-478 76720-480 76720-482 77081-796 77081-798 77081-800
tert-Butyl (1-(hydroxymethyl)cyclopropyl)carbamate 98%
tert-Butyl (1-(hydroxymethyl)cyclopropyl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₇NO₃
Boiling Pt: 294.4 °C (760 mm Hg)
Flash Pt: 131.88 °C
Storage Temperature: Ambient
MDL Number: MFCD09749954
CAS Number: 107017-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR