tert-Butyl ((5-bromopyrimidin-2-yl)methyl)carbamate ≥95%

Supplier: Ambeed
Warning

A297917-250MG A297917-100MG
103814-162EA 275 USD
103814-162 103835-306 77571-830 77612-712
tert-Butyl ((5-bromopyrimidin-2-yl)methyl)carbamate ≥95%
tert-Butyl ((5-bromopyrimidin-2-yl)methyl)carbamate

Precaución: Keep in a dark place, sealed in dry, at 2…8°C

Formula: C₁₀H₁₄BrN₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD15071123
CAS Number: 1235451-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR