tert-Butyl ((5-bromopyridin-2-yl)methyl)carbamate 97%

Supplier: Ambeed
Warning

A279761-100MG A279761-250MG A279761-5G
103813-478EA 71 USD
103813-478 76787-370 77668-236 77872-345
tert-Butyl ((5-bromopyridin-2-yl)methyl)carbamate 97%
tert-Butyl ((5-bromopyridin-2-yl)methyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD11520868
CAS Number: 1188477-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR