tert-Butyl ((1S,3S)-3-aminocyclopentyl)carbamate 95%

Supplier: Ambeed
Warning

A194852-5G A194852-1G A194852-250MG A194852-100MG
103816-698EA 120 USD
103816-698 76986-240 77085-246 77593-254 77611-868
tert-Butyl ((1S,3S)-3-aminocyclopentyl)carbamate 95%
tert-Butyl ((1S,3S)-3-aminocyclopentyl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11877864
CAS Number: 645400-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR