tert-Butyl ((1S,3R)-3-aminocyclohexyl)carbamate 95%

Supplier: Ambeed
Warning

A189556-1G A189556-5G A189556-250MG A189556-100MG
103816-200EA 193 USD
103816-200 103846-794 77084-564 77084-566 77164-180 77892-224
tert-Butyl ((1S,3R)-3-aminocyclohexyl)carbamate 95%
tert-Butyl ((1S,3R)-3-aminocyclohexyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD24469814
CAS Number: 1298101-47-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR