tert-Butyl ((1S,2S)-2-hydroxycyclohexyl)carbamate 97%

Supplier: Ambeed
Warning

A376843-10G A376843-250MG A376843-1G A376843-5G
103826-596EA 23 USD
103826-596 103837-622 76788-748 76788-750 76840-926 76840-928
tert-Butyl ((1S,2S)-2-hydroxycyclohexyl)carbamate 97%
tert-Butyl ((1S,2S)-2-hydroxycyclohexyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD11042330
CAS Number: 145166-06-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR