tert-Butyl ((1S,2S)-2-aminocyclopentyl)carbamate 95%

Supplier: Ambeed
Danger

A157078-250MG A157078-5G A157078-1G
103804-960EA 415 USD
103804-960 76871-900 76871-902 77318-976
tert-Butyl ((1S,2S)-2-aminocyclopentyl)carbamate 95%
tert-Butyl ((1S,2S)-2-aminocyclopentyl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Boiling Pt: 304.2±31.0 °C (760 mm Hg)
Flash Pt: 137.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11040590
CAS Number: 586961-34-4
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR