tert-Butyl ((1R,2R)-2-hydroxycyclopentyl)carbamate 95%

Supplier: Ambeed
Warning

A246909-250MG A246909-1G A246909-5G
103811-738EA 40 USD
103811-738 103851-572 76826-850 77304-398 77304-400
tert-Butyl ((1R,2R)-2-hydroxycyclopentyl)carbamate 95%
tert-Butyl ((1R,2R)-2-hydroxycyclopentyl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
MW: 201.26 g/mol
Melting Pt: 87.0 °C
Storage Temperature: Ambient
MDL Number: MFCD11656037
CAS Number: 454170-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR