tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate 97%
Supplier: Ambeed
|
Warning
|
A302560-100G
A302560-10G
A302560-1G
A302560-25G
A302560-5G
103814-344EA
8
USD
103814-344
103835-502
76723-632
76723-634
76838-180
76838-182
76838-184
tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate 97%
tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate
Precaución: for Research Use Only
Formula:
C₁₄H₂₂N₄O₄ Melting Pt: 86…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075122 CAS Number: 152120-54-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...