tert-Butoxybis(dimethylamino)methane 90%

Supplier: Ambeed
Danger

Synonyms: Bredereck's reagent, 1-tert-Butoxy-N,N,N',N'-tetramethylmethylenenediamine, [(tert-butoxy)(dimethylamino)methyl]dimethylamine, tert-Butoxy bis(dimethylamino)methane

A103181-10G A103181-25G A103181-1G A103181-5G
103796-500EA 5 USD
103796-500 103822-204 76687-120 76687-122 77283-052 77283-054
tert-Butoxybis(dimethylamino)methane 90%
tert-Butoxybis(dimethylamino)methane

Precaución: for Research Use Only

Formula: C₉H₂₂N₂O
MW: 174.288 g/mol
Boiling Pt: 50…55 °C (760 mm Hg)
Flash Pt: 41 °C
Storage Temperature: Freezer
MDL Number: MFCD00042858
CAS Number: 5815-08-7
UN: 2733
ADR: 3,8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR