Terephthalaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: Terephthaldialdehyde, Terephthaldicarboxaldehyde, p-Phthalaldehyde

A171486-100G A171486-500G A171486-1000G A171486-25G A171486-10G
103809-044EA 51 USD
103809-044 76784-238 76784-240 77301-134 77301-136 77550-106
Terephthalaldehyde 97%
Terephthalaldehyde

Precaución: for Research Use Only

Formula: C₈H₆O₂
MW: 134.13 g/mol
Boiling Pt: 246.5 °C (760 mm Hg)
Melting Pt: 114…116 °C
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00006949
CAS Number: 623-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR