Tenofovir disoproxil fumarate 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-(((((1-(6-Amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)phosphoryl)bis(oxy))bis(methylene) diisopropyl dicarbonate fumarate
A118576-100G
A118576-100MG
A118576-250MG
A118576-25G
A118576-500G
A118576-1G
A118576-5G
103799-078EA
10
USD
103799-078
76995-640
76995-642
76995-644
76995-646
76995-648
77311-746
77311-748
Tenofovir disoproxil fumarate 99%
Tenofovir disoproxil fumarate
Precaución: for Research Use Only
Formula:
C₁₉H₃₀N₅O₁₀P·C₄H₄O₄ MW: 635.5 g/mol Boiling Pt: 642.7 °C (760 mm Hg) Melting Pt: 113…115 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08141829 CAS Number: 202138-50-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...