Tenofovir 95%

Supplier: Ambeed
Danger

Synonyms: (R)-(((1-(6-Amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)phosphonic acid, (R)-[[2-(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl]phosphonic acid

A193687-100G A193687-10G A193687-500G A193687-5G A193687-1G A193687-50MG
103816-564EA 7 USD
103816-564 103847-178 76999-070 76999-072 76999-074 77085-076 77302-350 77593-244
Tenofovir 95%
Tenofovir

Precaución: for Research Use Only

Formula: C₉H₁₄N₅O₄P
MW: 287.22 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00943794
CAS Number: 147127-20-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR