4-((5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)carbamoyl)benzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Tamibarotene

A197861-100MG A197861-1MG A197861-250MG A197861-50MG A197861-5G
103792-364EA 11 USD
103792-364 103799-468 103816-972 76691-104 76691-106 76691-108 77085-608 77625-978
4-((5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)carbamoyl)benzoic acid 98%
4-((5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)carbamoyl)benzoic acid

Precaución: for Research Use Only

Formula: C₂₂H₂₅NO₃
MW: 351.45 g/mol
Boiling Pt: 449.6 °C (760 mm Hg)
Melting Pt: 231…232 °C
Storage Temperature: Freezer
MDL Number: MFCD00866188
CAS Number: 94497-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR