Tadalafil 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(6R,12aR)-6-(Benzo[d][1,3]dioxol-5-yl)-2-methyl-2,3,12,12a-tetrahydropyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4(6H,7H)-dione
A147960-1000G
A147960-100G
A147960-100MG
A147960-10G
A147960-500G
A147960-1G
A147960-5G
A147960-25G
103803-854EA
9
USD
103803-854
103841-970
77110-114
77110-116
77110-118
77110-120
77110-122
77299-618
77299-620
77317-394
Tadalafil 98%
Tadalafil
Precaución: for Research Use Only
Formula:
C₂₂H₁₉N₃O₄ MW: 389.4 g/mol Boiling Pt: 679.1 °C (760 mm Hg) Melting Pt: 298…300 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07771966 CAS Number: 171596-29-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...