Diphenyl sulfoxide 98%

Supplier: Ambeed
Warning

Synonyms: Phenyl sulphoxide, sulphinyldibenzene

A116394-100G A116394-10G A116394-25G A116394-500G A116394-5G
103808-154EA 117 USD
103808-154 103824-588 76797-082 76797-084 76797-086 76797-088 77297-460
Diphenyl sulfoxide 98%
Diphenyl sulphoxide

Precaución: for Research Use Only

Formula: C₁₂H₁₀OS
MW: 202.28 g/mol
Boiling Pt: 357.6 °C (760 mm Hg)
Melting Pt: 66…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00002085
CAS Number: 945-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR