Sulfamethoxazole 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-N-(5-methylisoxazol-3-yl)benzenesulphonamide

A205081-25G A205081-1000G A205081-100G A205081-500G A205081-5G
103848-258EA 5 USD
103848-258 77086-458 77139-452 77139-454 77139-456 77139-458
Sulfamethoxazole 98%
Sulfamethoxazole

Precaución: for Research Use Only

Formula: C₁₀H₁₁N₃O₃S
MW: 253.28 g/mol
Melting Pt: 166 °C
Flash Pt: 245.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00010546
CAS Number: 723-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR