Strontium-2,2'-((5-carboxylato-4-(carboxylatomethyl)-3-cyanothiophen-2-yl)azanediyl)diacetate 98%

Supplier: Ambeed
Warning

Synonyms: 2,2'-((5-Carboxy-4-(carboxymethyl)-3-cyanothiophen-2-yl)azanediyl)diacetic acid, distrontium salt, Strontium Ranelate

A162052-100G A162052-100MG A162052-10G A162052-10MG A162052-50MG A162052-25G A162052-500G
103843-810EA 18 USD
103843-810 76783-896 76783-898 76783-900 76783-902 76783-904 77300-570 77667-268
Strontium-2,2'-((5-carboxylato-4-(carboxylatomethyl)-3-cyanothiophen-2-yl)azanediyl)diacetate 98%
Strontium-2,2'-((5-carboxylato-4-(carboxylatomethyl)-3-cyanothiophen-2-yl)azanediyl)diacetate

Precaución: for Research Use Only

Formula: C₁₂H₆N₂O₈SSr₂
Storage Temperature: Freezer
MDL Number: MFCD09038742
CAS Number: 135459-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR