Spiro[indoline-3,4'-piperidin]-2-one 97%

Supplier: Ambeed
Warning

A184313-1G A184313-5G
103815-742EA 119 USD
103815-742 103846-304 77083-918 77083-920
Spiro[indoline-3,4'-piperidin]-2-one 97%
Spiro[indoline-3,4'-piperidin]-2-one

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O
MW: 202.2500 g/mol
Flash Pt: 181.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12198581
CAS Number: 252882-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR