4-Spiroindenepiperidine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Spiro[indene-1,4'-piperidine] hydrochloride

A101379-250MG A101379-1G
103795-942EA 82 USD
103795-942 103821-626 76970-164 77297-078
4-Spiroindenepiperidine hydrochloride 95%
4-Spiroindenepiperidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₁₆ClN
MW: 221.73 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01632538
CAS Number: 137730-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR