Spiro[cyclopropane-1,3'-indolin]-2'-one 95%

Supplier: Ambeed
Warning

A270978-100MG A270978-250MG A270978-1G
103813-022EA 133 USD
103813-022 103852-930 77014-418 77014-420 77092-702
Spiro[cyclopropane-1,3'-indolin]-2'-one 95%
Spiro[cyclopropane-1,3'-indolin]-2'-one

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MW: 159.19 g/mol
Boiling Pt: 356.8 °C (760 mm Hg)
Flash Pt: 207.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11934419
CAS Number: 13861-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR