Spiro[2.5]octan-6-ylmethanol 97%

Supplier: Ambeed
Warning

A246139-100MG A246139-250MG
103811-690EA 273 USD
103811-690 76874-508 77612-372
Spiro[2.5]octan-6-ylmethanol 97%
Spiro[2.5]octan-6-ylmethanol

Precaución: for Research Use Only

Formula: C₉H₁₆O
MW: 140.223 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11501505
CAS Number: 849671-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR