Spermine tetrahydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: N1,N1'-(Butane-1,4-diyl)bis(propane-1,3-diamine) tetrahydrochloride

A134234-10G A134234-100G A134234-5G A134234-1G A134234-25G
103801-988EA 13 USD
103801-988 76719-068 76782-546 77298-700 77314-716 77314-718
Spermine tetrahydrochloride 95%
Spermine tetrahydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₃₀Cl₄N₄
Boiling Pt: 308.4 °C (760 mm Hg)
Flash Pt: 175.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00012914
CAS Number: 306-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR