Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Kobayashi's Reagent, NaBArF24
A136880-100G
A136880-10G
A136880-250MG
A136880-1G
A136880-25G
A136880-5G
103802-368EA
15
USD
103802-368
76719-224
76719-226
76719-228
77315-286
77315-288
77315-290
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate 97%
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate
Precaución: for Research Use Only
Formula:
C₃₂H₁₂BF₂₄Na MW: 886.20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00043323 CAS Number: 79060-88-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...