Sodium toluene-4-sulfonate 90%

Supplier: Ambeed
Warning

A752002-1000G A752002-100G A752002-500G A752002-25G
103840-856EA 5 USD
103840-856 76728-132 76728-134 76728-136 77308-256
Sodium toluene-4-sulfonate 90%
Sodium toluene-4-sulphonate

Precaución: for Research Use Only

Formula: C₇H₇NaO₃S
Storage Temperature: Ambient
MDL Number: MFCD00798566
CAS Number: 657-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR