Sodium valerate 97%

Supplier: Ambeed
Warning

Synonyms: Sodium pentanoate

A237709-25G A237709-5G A237709-10G A237709-250MG A237709-500G A237709-100G
103811-172EA 6 USD
103811-172 103850-954 77090-052 77090-054 77165-688 77165-690 77165-692 77568-300
Sodium valerate 97%
Sodium valerate

Precaución: For research use only.

Formula: C₅H₉NaO₂
MW: 124.12 g/mol
Flash Pt: 80.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00066205
CAS Number: 6106-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR