Ethane sulfinic acid sodium salt 95%

Supplier: Ambeed
Warning

A122844-100G A122844-10G A122844-25G A122844-1G A122844-5G
103800-290EA 12 USD
103800-290 76797-476 76797-478 76797-480 77312-410 77312-412
Ethane sulfinic acid sodium salt 95%
Ethane sulfinic acid sodium salt

Precaución: for Research Use Only

Formula: C₂H₅NaO₂S
Boiling Pt: 244 °C (760 mm Hg)
Melting Pt: 292 °C
Flash Pt: 101.4 °C
Storage Temperature: Ambient
MDL Number: MFCD06659476
CAS Number: 20035-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR