Disodium-1,4-butanedisulfonate 98%

Supplier: Ambeed
Warning

Synonyms: 1,4-Butanedisulphonic Acid Disodium Salt, Sodium butane-1,4-disulphonate

A236312-5G A236312-1G
103850-848EA 9 USD
103850-848 77089-896 77193-230
Disodium-1,4-butanedisulfonate 98%
Disodium-1,4-butanedisulfonate

Precaución: for Research Use Only

Formula: C₄H₁₀O₆S₂
MW: 262.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059839
CAS Number: 36589-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR