Pyranine 85%

Supplier: Ambeed
Warning

A218894-100G A218894-10G A218894-5G A218894-500G
103849-626EA 8 USD
103849-626 76721-904 76721-906 77088-166 77667-832
Pyranine 85%
Pyranine

Precaución: for Research Use Only

Formula: C₁₆H₇Na₃O₁₀S₃
MW: 524.371 g/mol
Melting Pt: >300 °C
Flash Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD00037575
CAS Number: 6358-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR