Sodium 8-(2-hydroxybenzamido)octanoate 97%

Supplier: Ambeed
Warning

Synonyms: Salcaprozate sodium

A224096-1G A224096-5G A224096-100G A224096-250MG A224096-25G A224096-50MG
103819-084EA 23 USD
103819-084 103849-974 77088-608 77088-610 77098-012 77098-014 77098-016 77685-607
Sodium 8-(2-hydroxybenzamido)octanoate 97%
Sodium 8-(2-hydroxybenzamido)octanoate

Precaución: for Research Use Only

Formula: C₁₅H₂₀NNaO₄
MW: 301.31 g/mol
Boiling Pt: 521.7 °C (760 mm Hg)
Flash Pt: 269.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00952954
CAS Number: 203787-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR