Sodium 4-phenylbutyrate 98%

Supplier: Ambeed
Warning

A455726-100MG A455726-1MG A455726-250MG A455726-100G
103827-844EA 22 USD
103827-844 103838-918 76725-628 76725-630 76725-632 76725-634 77872-750
Sodium 4-phenylbutyrate 98%
Sodium 4-phenylbutyrate

Precaución: for Research Use Only

Formula: C₁₀H₁₁O₂·Na
Boiling Pt: 272.9 °C (760 mm Hg)
Flash Pt: 170.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00800247
CAS Number: 1716-12-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR