p-Toluenesulfinic Acid Sodium Salt 97%

Supplier: Ambeed
Warning

A288742-100G A288742-500G A288742-1000G A288742-25G
103809-962EA 57 USD
103809-962 76723-448 76723-450 77305-540 77305-542
p-Toluenesulfinic Acid Sodium Salt 97%
p-Toluenesulfinic Acid Sodium Salt

Precaución: for Research Use Only

Formula: C₇H₇NaO₂S
Melting Pt: >300 °C
Flash Pt: 159.4 °C
Storage Temperature: Freezer
MDL Number: MFCD00013136
CAS Number: 824-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR