4-Fluorobenzenesulfinic acid sodium salt 95%

Supplier: Ambeed
Warning

A204921-25G A204921-100G A204921-10G A204921-500G A204921-1G A204921-5G
103817-578EA 8 USD
103817-578 103848-244 77086-442 77287-770 77287-772 77287-774 77302-920 77302-922
4-Fluorobenzenesulfinic acid sodium salt 95%
4-Fluorobenzenesulfinic acid sodium salt

Precaución: for Research Use Only

Formula: C₆H₄FO₂S·Na
Melting Pt: >250 °C
Flash Pt: 153.1 °C
Storage Temperature: Freezer
MDL Number: MFCD03093775
CAS Number: 824-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR