Sodium 4-(trifluoromethyl)benzoate 97%

Supplier: Ambeed
Warning

A145316-100G A145316-10G A145316-25G A145316-5G
103841-604EA 7 USD
103841-604 77095-594 77095-596 77299-446 77316-888
Sodium 4-(trifluoromethyl)benzoate 97%
Sodium 4-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₈H₄F₃NaO₂
MW: 212.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458184
CAS Number: 25832-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR