Sodium-3-methyl-2-oxobutanoate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Methyl-2-oxobutanoic acid, sodium salt, Sodium 3-methyl-2-oxobutanoate
A232192-1G
A232192-5G
A232192-250MG
103810-820EA
33
USD
103810-820
103850-572
77089-494
77089-496
77275-768
Sodium-3-methyl-2-oxobutanoate 95%
Sodium-3-methyl-2-oxobutanoate
Precaución: for Research Use Only
Formula:
C₅H₇NaO₃ MW: 138.098 g/mol Boiling Pt: 170.2 °C (760 mm Hg) Melting Pt: 227…230 °C at N/A mmHg Flash Pt: 71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002581 CAS Number: 3715-29-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...