Triphenylphosphine-3,3',3''-trisulfonic acid trisodium salt hydrate 85%

Supplier: Ambeed
Warning

Synonyms: Sodium 3,3',3''-phosphinetriyltribenzenesulphonate, Trisodium 3,3',3''-Phosphinetriyltribenzenesulphonate, Tris(3-sulphophenyl)phosphine trisodium salt, Tris(3-sulphonatophenyl)phosphine, sodium salt

A189888-100G A189888-1G A189888-25G A189888-5G
103816-230EA 8 USD
103816-230 103846-826 76784-874 77084-598 77302-190 77302-192
Triphenylphosphine-3,3',3''-trisulfonic acid trisodium salt hydrate 85%
Triphenylphosphine-3,3',3''-trisulfonic acid trisodium salt hydrate

Precaución: for Research Use Only

Formula: C₁₈H₁₂Na₃O₉PS₃
Storage Temperature: Ambient
MDL Number: MFCD00145472
CAS Number: 63995-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR