CAPS sodium salt (3-(cyclohexylamino)propanesulfonic acid sodium salt) 98%

Supplier: Ambeed
Warning

Synonyms: Sodium 3-(cyclohexylamino)propane-1-sulphonate

A826855-100G A826855-25G A826855-5G
103841-158EA 8 USD
103841-158 76698-210 76845-392 76845-394
CAPS sodium salt (3-(cyclohexylamino)propanesulfonic acid sodium salt) 98%
CAPS sodium salt (3-(Cyclohexylamino)propanesulphonic acid sodium salt)

Precaución: for Research Use Only

Formula: C₉H₁₈NNaO₃S
MW: 243.3000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00269798
CAS Number: 105140-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR