Sodium 2-phenyl-1H-indole-5-sulfonate 95%

Supplier: Ambeed
Warning

A140369-10G A140369-5G
76996-844EA 12.74 USD
76996-844 77315-976
Sodium 2-phenyl-1H-indole-5-sulfonate 95%
Sodium 2-phenyl-1H-indole-5-sulfonate

Precaución: for Research Use Only

Formula: C₁₄H₁₀NNaO₃S
MW: 295.29 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04972100
CAS Number: 119205-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR