Sodium-2-naphthalenesulfonate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Naphthalenesulphonic acid Sodium Salt, Sodium 2-naphthalenesulphonate
A495467-25G
A495467-5G
A495467-100G
103839-332EA
5
USD
103839-332
76843-298
76843-300
77279-656
Sodium-2-naphthalenesulfonate 97%
Sodium-2-naphthalenesulfonate
Precaución: for Research Use Only
Formula:
C₁₀H₈O₃S MW: 230.21 g/mol Melting Pt: 275 °C at N/A mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00064186 CAS Number: 532-02-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...