Sodium chlorodifluoroacetate 97%
Supplier: Ambeed
|
Warning
|
A280716-25G
A280716-5G
A280716-100G
A280716-10G
A280716-500G
103853-464EA
5
USD
103853-464
76837-150
76837-152
77114-048
77114-050
77114-052
Sodium chlorodifluoroacetate 97%
Sodium chlorodifluoroacetate
Precaución: for Research Use Only
Formula:
C₂ClF₂NaO₂ MW: 152.46 g/mol Melting Pt: 196…198 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064771 CAS Number: 1895-39-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...