Disodium-4,4'-bis(2-sulfonatostyryl)biphenyl 97%

Supplier: Ambeed
Warning

Synonyms: Sodium 2,2'-([1,1'-biphenyl]-4,4'-diylbis(ethene-2,1-diyl))dibenzenesulphonate, 4,4'-Bis(2-sulphonatostyryl)biphenyl Disodium Salt

A442962-25G A442962-10G
103810-342EA 107 USD
103810-342 76842-534 77291-620
Disodium-4,4'-bis(2-sulfonatostyryl)biphenyl 97%
Disodium-4,4'-bis(2-sulfonatostyryl)biphenyl

Precaución: for Research Use Only

Formula: C₂₈H₂₂O₆S₂
MW: 562.56 g/mol
Melting Pt: >320 °C
Storage Temperature: Ambient
MDL Number: MFCD00318874
CAS Number: 27344-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR